Anti-ESR1 Mouse monoclonal antibody [clone: 33]

ABCAAB2746-50EA 819 CHF
ABCAAB2746-50
Anti-ESR1 Mouse monoclonal antibody [clone: 33]
Type: Primary
Antigen: ESR1 (estrogen receptor 1)
Clonality: monoclonal
Clone: 33
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR