Anti-LAP1 Mouse Monoclonal Antibody [clone: RL13]
Lieferant: Abcam
ABCAAB2737-200EA 505.57 CHF
ABCAAB2737-200
Anti-LAP1 Mouse Monoclonal Antibody [clone: RL13]
Antikörper
Type: Primary
Antigen: LAP1
Clonality: monoclonal
Clone: RL13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat