Anti-PKD1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB2634-100EA 911 CHF
ABCAAB2634-100
Anti-PKD1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PKD1 (polycystic kidney disease 1 (autosomal dominant))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR