Anti-EIF4EBP1 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB2606-50EA 539.48 CHF
ABCAAB2606-50
Anti-EIF4EBP1 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR