Anti-Hepatitis C Virus NS5a Rabbit polyclonal antibody

ABCAAB2594-100EA 732 CHF
ABCAAB2594-100
Anti-Hepatitis C Virus NS5a Rabbit polyclonal antibody
Type: Primary
Antigen: Hepatitis C Virus NS5a
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR