Anti-Alpha Fodrin Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB2558-50EA 932 CHF
ABCAAB2558-50
Anti-Alpha Fodrin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Alpha Fodrin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR