Anti-CASP6 Rabbit polyclonal antibody

ABCAAB2552-50EA 744 CHF
ABCAAB2552-50
Anti-CASP6 Rabbit polyclonal antibody
Type: Primary
Antigen: CASP6 (caspase 6, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR