Anti-CAPN1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB2543-50EA 911 CHF
ABCAAB2543-50
Anti-CAPN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CAPN1 (calpain 1, (mu/I) large subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR