Anti-KRT8 Mouse Monoclonal Antibody [clone: C-43]

Supplier: Abcam
ABCAAB2530-100EA 529.11 CHF
ABCAAB2530-100
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-43]
Antibodies
Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone: C-43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR