Anti-CD98 Mouse Monoclonal Antibody [clone: MEM-108]

Supplier: Abcam
ABCAAB2528-100EA 508.36 CHF
ABCAAB2528-100
Anti-CD98 Mouse Monoclonal Antibody [clone: MEM-108]
Antibodies
Type: Primary
Antigen: CD98
Clonality: monoclonal
Clone: MEM-108
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR