Anti-Heparan Sulphate Proteoglycan (Large) Rat Monoclonal Antibody [clone: A7L6]

Lieferant: Abcam
ABCAAB2501-50EA 548.5 CHF
ABCAAB2501-50
Anti-Heparan Sulphate Proteoglycan (Large) Rat Monoclonal Antibody [clone: A7L6]
Antikörper
Type: Primary
Antigen: Heparan Sulphate Proteoglycan (Large)
Clonality: monoclonal
Clone: A7L6
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR