Anti-FASLG Rabbit polyclonal antibody

ABCAAB2440-1EA 708 CHF
ABCAAB2440-1
Anti-FASLG Rabbit polyclonal antibody
Type: Primary
Antigen: FASLG (Fas ligand (TNF superfamily, member 6))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR