Anti-FAS Rabbit polyclonal antibody

ABCAAB2437-1EA 716 CHF
ABCAAB2437-1
Anti-FAS Rabbit polyclonal antibody
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR