Anti-BRCA1 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB2401-50EA 920 CHF
ABCAAB2401-50
Anti-BRCA1 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR