Anti-BID Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB2388-50EA 492.8 CHF
ABCAAB2388-50
Anti-BID Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR