Anti-MCM7 Mouse Monoclonal Antibody [clone: 47DC141]

Supplier: Abcam
ABCAAB2360-500EA 549.86 CHF
ABCAAB2360-500
Anti-MCM7 Mouse Monoclonal Antibody [clone: 47DC141]
Antibodies
Type: Primary
Antigen: MCM7 (minichromosome maintenance complex component 7)
Clonality: monoclonal
Clone: 47DC141
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR