Anti-ERCC1 Mouse Monoclonal Antibody [clone: 8F1]
Lieferant: Abcam
ABCAAB2356-500EA 548.5 CHF
ABCAAB2356-500
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 8F1]
Antikörper
Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: monoclonal
Clone: 8F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat