Anti-ANXA10 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB2343-100EA 910 CHF
ABCAAB2343-100
Anti-ANXA10 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ANXA10 (annexin A10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR