Anti-Prostaglandin E2 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB2318-100EA 0 CHF
ABCAAB2318-100
Anti-Prostaglandin E2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Prostaglandin E2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now