Anti-CASP3 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB2302-50EA 586.17 CHF
ABCAAB2302-50
Anti-CASP3 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR