Anti-COP Goat polyclonal antibody

ABCAAB2225-100EA 458 CHF
ABCAAB2225-100
Anti-COP Goat polyclonal antibody
Type: Primary
Antigen: COP (clathrin-ordered protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR