Anti-Prostate Specific Antigen Mouse monoclonal antibody [clone: A67-B/E3]

Supplier: Abcam
ABCAAB2218-100EA 508.36 CHF
ABCAAB2218-100
Anti-Prostate Specific Antigen Mouse monoclonal antibody [clone: A67-B/E3]
Type: Primary
Antigen: Prostate Specific Antigen
Clonality: monoclonal
Clone: A67-B/E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR