Anti-ZBP1 Sheep Polyclonal Antibody

Lieferant: Abcam
ABCAAB2079-125EA 0 CHF
ABCAAB2079-125
Anti-ZBP1 Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZBP1 (Z-DNA binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR