Anti-ZIP Kinase Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB2057-100EA 518.74 CHF
ABCAAB2057-100
Anti-ZIP Kinase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZIP Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR