Anti-Hepatitis B Virus Surface Antigen Mouse monoclonal antibody [clone: NE3]

ABCAAB2044-500EA 672 CHF
ABCAAB2044-500
Anti-Hepatitis B Virus Surface Antigen Mouse monoclonal antibody [clone: NE3]
Type: Primary
Antigen: Hepatitis B Virus Surface Antigen
Clonality: monoclonal
Clone: NE3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR