Anti-DAXX Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB2017-100EA 524.65 CHF
ABCAAB2017-100
Anti-DAXX Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DAXX (death-domain associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR