Anti-TG Mouse monoclonal antibody [clone: 12G9]

ABCAAB1983-500EA 737 CHF
ABCAAB1983-500
Anti-TG Mouse monoclonal antibody [clone: 12G9]
Type: Primary
Antigen: TG (thyroglobulin)
Clonality: monoclonal
Clone: 12G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR