Anti-INS Mouse Monoclonal Antibody [clone: 3A6]

Supplier: Abcam
ABCAAB1965-100EA 911 CHF
ABCAAB1965-100
Anti-INS Mouse Monoclonal Antibody [clone: 3A6]
Antibodies
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone: 3A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR