Anti-CGB5 Mouse monoclonal antibody [clone: G9]

ABCAAB1958-500EA 699 CHF
ABCAAB1958-500
Anti-CGB5 Mouse monoclonal antibody [clone: G9]
Type: Primary
Antigen: CGB5 (chorionic gonadotropin, beta polypeptide 5)
Clonality: monoclonal
Clone: G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR