Anti-IgG1 Mouse Monoclonal Antibody

Supplier: Abcam
ABCAAB1927-1EA 482.42 CHF
ABCAAB1927-1
Anti-IgG1 Mouse Monoclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR