Anti-APOE Mouse monoclonal antibody unconjugated [clone: D6E10]
Lieferant: Abcam
ABCAAB1906-100EA 548.5 CHF
ABCAAB1906-100
Anti-APOE Mouse monoclonal antibody unconjugated [clone: D6E10]
Antikörper
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: monoclonal
Clone: D6E10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse