Anti-AGTR1 Sheep polyclonal antibody

ABCAAB1886-125EA 660 CHF
ABCAAB1886-125
Anti-AGTR1 Sheep polyclonal antibody
Type: Primary
Antigen: AGTR1 (angiotensin II receptor, type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR