Anti-CAT Rabbit polyclonal antibody

ABCAAB1877-10EA 741 CHF
ABCAAB1877-10
Anti-CAT Rabbit polyclonal antibody
Type: Primary
Antigen: CAT (catalase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR