Anti-Liver Carboxylesterase 1 Rabbit polyclonal antibody

ABCAAB1875-1EA 672 CHF
ABCAAB1875-1
Anti-Liver Carboxylesterase 1 Rabbit polyclonal antibody
Type: Primary
Antigen: Liver Carboxylesterase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR