Anti-VSV-G Tag Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB1874-100EA 508.36 CHF
ABCAAB1874-100
Anti-VSV-G Tag Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VSV-G Tag (VSV-G tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR