Anti-Caldesmon Mouse monoclonal antibody [clone: h-CALD]

ABCAAB1826-500EA 741 CHF
ABCAAB1826-500
Anti-Caldesmon Mouse monoclonal antibody [clone: h-CALD]
Type: Primary
Antigen: Caldesmon
Clonality: monoclonal
Clone: h-CALD
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR