Anti-HA tag Mouse monoclonal antibody FITC (Fluorescein) [clone: 2C16]

ABCAAB1789-50EA 737 CHF
ABCAAB1789-50
Anti-HA tag Mouse monoclonal antibody FITC (Fluorescein) [clone: 2C16]
Type: Primary
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: 2C16
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR