Anti-CCR4 Rabbit polyclonal antibody

ABCAAB1679-50EA 699 CHF
ABCAAB1679-50
Anti-CCR4 Rabbit polyclonal antibody
Type: Primary
Antigen: CCR4 (chemokine (C-C motif) receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR