Anti-TNFRSF10B Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB1675-100EA 920 CHF
ABCAAB1675-100
Anti-TNFRSF10B Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR