Anti-CCR5 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB1673-100EA 941 CHF
ABCAAB1673-100
Anti-CCR5 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR