Anti-CCR8 Goat polyclonal antibody

ABCAAB1665-100EA 660 CHF
ABCAAB1665-100
Anti-CCR8 Goat polyclonal antibody
Type: Primary
Antigen: CCR8 (chemokine (C-C motif) receptor 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR