Anti-CCR6 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB1659-100EA 933 CHF
ABCAAB1659-100
Anti-CCR6 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCR6 (chemokine (C-C motif) receptor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR