Anti-HDAC6 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB1440-50EA 519.88 CHF
ABCAAB1440-50
Anti-HDAC6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HDAC6 (histone deacetylase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR