Anti-BAD Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB1435-50EA 477.24 CHF
ABCAAB1435-50
Anti-BAD Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: BAD (BCL2-associated agonist of cell death)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR