Anti-TERF1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB1423-100EA 457.88 CHF
ABCAAB1423-100
Anti-TERF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TERF1 (telomeric repeat binding factor (NIMA-interacting) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse