Anti-KRT10 Mouse monoclonal antibody [clone: VIK-10]

ABCAAB1421-100EA 660 CHF
ABCAAB1421-100
Anti-KRT10 Mouse monoclonal antibody [clone: VIK-10]
Type: Primary
Antigen: KRT10 (keratin 10)
Clonality: monoclonal
Clone: VIK-10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR