Anti-CD58 Mouse monoclonal antibody [clone: MEM-63]

ABCAAB1420-100EA 660 CHF
ABCAAB1420-100
Anti-CD58 Mouse monoclonal antibody [clone: MEM-63]
Type: Primary
Antigen: CD58 (CD58 molecule)
Clonality: monoclonal
Clone: MEM-63
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR