Anti-CDH1 Mouse Monoclonal Antibody [clone: HECD-1]

Supplier: Abcam
ABCAAB1416-250EA 596.55 CHF
ABCAAB1416-250
Anti-CDH1 Mouse Monoclonal Antibody [clone: HECD-1]
Antibodies
Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: monoclonal
Clone: HECD-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR