Anti-Myc Tag Chicken polyclonal antibody FITC (Fluorescein)

ABCAAB1394-100EA 720 CHF
ABCAAB1394-100
Anti-Myc Tag Chicken polyclonal antibody FITC (Fluorescein)
Type: Primary
Antigen: Myc Tag (Myc tag)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Chicken
Isotype: unknown
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR