Anti-SEC61A Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB1327-100EA 524.65 CHF
ABCAAB1327-100
Anti-SEC61A Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEC61A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR